Check Order Status
Spatula with Stainless-Steel Blade
Actual product may vary
Spatula with Stainless-Steel Blade
Fisherbrand Spatula with Stainless-Steel Blade are non-stick, corrosion-free with a bright finish.
Have Questions?
Catalog Number
Length (Metric) Blade
FIS14-365A
7.62 cm
Catalog number
FIS14-365A
Price (SGD)
38.00
EA
Add to cart
Length (Metric) Blade:
7.62 cm
Price (SGD)
38.00
EA
Add to cart
Fisherbrand™ Spatula with Stainless-Steel Blade are non-stick, corrosion-free with a bright finish
Multipurpose - Useful in educational, research or healthcare applications
Multiple sizes allow for varied laboratory applications
Made from ASTM stainless steel
Non-sticking, corrosion-free standard blade
Riveted, smooth hardwood handle designed and engineered to fit comfortably in your hand
Electronically passivated for corrosion resistance
Autoclavable
Latex-free
Specifications
Product Type
Spatula
Finish
Bright
Length (English) Blade
3 in.
Length (Metric) Blade
7.62 cm
Blade Material
Stainless Steel
Handle Material
Hardwood
Handle Type
Riveted