Search Fisher Scientific
| Catalog Number | Sieve Number | Pore Size |
|---|---|---|
| FIS14-294-88 | 140 | 106 μm |
| FIS14-294-87 | 120 | 125 μm |
| FIS14-294-71 | 10 | 2 mm |
| FIS14-294-79 | 35 | 500 μm |
| FIS14-294-84 | 70 | 212 μm |
For use with Advantech Model L3P Sonic Sifters
Meet all applicable ASTM™ standards. 3 in. I.D. (7.6cm) sieves with clear acrylic frames. Choice of stainless-steel or nickel-mesh sieves.
Standard Stainless-steel Mesh Sieves
Precision Nickel Mesh Sieves