Stomacher™ Lab Blender Bags
Actual product may vary
Seward
Stomacher™ Lab Blender Bags
The Stomacher™ Bag range has been designed to fulfill all applications from sample blending, straining and storage, including bags designed for extensive blending and small samples for life science applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)DescriptionNo. per Pack
FIS14-285-980 to 400 mLSquare bottom, Closure250
FIS14-285-2580 to 400 mLSquare bottom, Standard500
FIS14-285-2480 to 400 mLRound bottom, Standard500
FKS1074685580 to 400 mLRound bottom; with Strainer200
FKS1065816580 to 400 mLSquare bottom, With strainer200
FIS14-285-2080 to 400 mLRound bottom; Closure250
FKS1054739480 to 400 mLRound bottom; Closure250
Catalog number FIS14-285-9
Price (SGD)
839.00
Case
Capacity (Metric):
80 to 400 mL
Description:
Square bottom, Closure
No. per Pack:
250
Price (SGD)
839.00
Case
  • Seward Stomacher bag range is made from high-quality food-grade virgin LDPE granule extruded to 65μm film thickness
  • Heavy-duty bags also available for large volume sampling 80μm film thickness; Sterile (gamma-irradiated) polyethylene bags; Closure bags feature wire closure strip across bag top for short term storage.
  • Strainer bags have inner plastic mesh to make it easy to pipette from samples that produce a large amount of debris
  • Single-chamber bags for use in Lab Blenders; internal sterility assured with proprietary tear-off sterile barrier top on sachets
  • Highly transparent and puncture-resistant virgin polyethylene extruded tube material
  • Simply tear the perforated end along the top of the sachet, remove the bag and open using Seward bag opener, insert the material to be analyzed and processed
  • Bags available for sample size 25ul - 4500ml
Specifications
DescriptionSquare bottom, Closure
AutoclavableNot Autoclavable
Closure TypeFlat Wire
ColorClear
DisposableSingle-use
For Use With (Application)To prepare, store and transport samples in microbiology sample preparation
For Use With (Equipment)Stomacher 400 and all other 400ml paddle blenders
MaterialPolyethylene
SterilitySterile
No. per Pack250
Sterilization MethodGamma Irradiated
Dimensions (L x W)305mm x 177mm
Product TypeLab Blender Bags
Capacity (Metric)80 to 400 mL
Length (Metric)305 mm
Width (Metric)177 mm
Thickness (Metric)65 μm
Unit SizeCase