Search Thermo Fisher Scientific
| Catalog Number | Product Type | Holds | For Use With (Equipment) |
|---|---|---|---|
| FIS14-272-4AA | Flask Clamp | 125 mL Erlenmeyer Flask | Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers |
Mix and match Thermo Scientific™ Solaris™ shaker Universal clamps to suit individual needs in the laboratory.