Specifications
Product TypeLadle
MaterialHDPE
Capacity (English)20.3 oz.
Capacity (Metric)600 mL
For Use With (Application)For sampling and dispensing corrosive liquid
Dimensions (L x W x H)5.03 x 4 x 13 in. (12.77 x 10.16 x 33.02 cm)
Height (English)13 in.
Height (Metric)33.02 cm
Length (English)5.03 in.
Length (English) Handle10 in.
Length (Metric)12.77 cm
Length (Metric) Handle25.4 cm
Width (English)4 in.
Width (Metric)10.16 cm
Unit SizeEach