Reusable Polyethylene Dippers
Actual product may vary
Reusable Polyethylene Dippers
Reusable Polyethylene Dippers
Reusable Polyethylene Dippers
500mL beakers with long handles for removing samples from large tanks.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric) HandleLength (English) Handle
FIS14-242-103.7 m12 ft.
FIS14-242-10.9 m3 ft.
FIS14-242-51.8 m6 ft.
Catalog number FIS14-242-10
Price (SGD)
350.00
Each
Length (Metric) Handle:
3.7 m
Length (English) Handle:
12 ft.
Price (SGD)
350.00
Each
500mL beakers with long handles for removing samples from large tanks.
  • Lightweight, durable polyethylene
  • Graduated beaker features dual pouring spouts for easy left- or right-handed operation
  • Handle has an “easy-grip” end for secure handling under adverse conditions
Specifications
Product TypeDipper
MaterialHDPE
Capacity (Metric)500 mL
For Use With (Application)For removing samples from large tanks
Dimensions (L x W x H)150.5 x 5 x 4.25 in. (382.2 x 12.7 x 10.79 cm)
Height (English)4.25 in.
Height (Metric)10.79 cm
Length (English)150.5 in.
Length (English) Handle12 ft.
Length (Metric)382.2 cm
Length (Metric) Handle3.7 m
Width (English)5 in.
Width (Metric)12.7 cm
Unit SizeEach