0.2mL PCR Tube Strips
Actual product may vary
Fisherbrand
0.2mL PCR Tube Strips
Ideal for use in 0.2mL, 96-well V-bottom thermal cyclers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorClosure Type
FIS14-230-215NaturalFlat Cap
FIS14-230-210NaturalDomed Cap
FKS12179770NaturalFlat Cap
FIS14-230-216RedFlat Cap
FIS14-230-217BlueFlat Cap
FIS14-230-218GreenFlat Cap
FIS14-230-219YellowFlat Cap
Catalog number FIS14-230-215
Price (SGD)
336.00
Case
Color:
Natural
Closure Type:
Flat Cap
Price (SGD)
336.00
Case
Ideal for use in 0.2mL, 96-well V-bottom thermal cyclers
  • 0.25mL maximum tube volume when capped
  • Strips of eight tubes
  • Can be cut into individual tubes
  • Thin-wall design
  • Cleanroom produced
  • Virgin polypropylene
  • Available with flat cap strips, domed cap strips, or without cap strips
Specifications
Certifications/ComplianceHuman DNA, RNase, DNase, endotoxins free
ColorNatural
Volume (Metric)0.2 mL
Quantity250 Case
For Use With (Equipment)0.2mL 96-well V-bottom thermal cyclers, PCR and qPCR/Real-Time PCR applications
MaterialPolypropylene
Closure TypeFlat Cap
No. of Wells96
FormatStrip
Product TypeThin Wall 8-Strip PCR Tube
Unit SizeCase