250 μL Automation Tips for Agilent™/Velocity 11 VPrep™ and Bravo, 96 Tip Format
Actual product may vary
250 μL Automation Tips for Agilent™/Velocity 11 VPrep™ and Bravo, 96 Tip Format
For use with Agilent/Velocity11 automation systems
Have Questions?
Catalog NumberSterilityTip StyleFeatures
FIS14-222-881Non-sterileStandardNon-pyrogenic
Catalog number FIS14-222-881
Price (SGD)
230.00
Pack
Sterility:
Non-sterile
Tip Style:
Standard
Features:
Non-pyrogenic
Price (SGD)
230.00
Pack
Tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality.
  • Clear polypropylene
  • Maxymum Recovery™ improves accuracy and precision while reducing sample loss
  • Certified RNase-/DNase-free and nonpyrogenic
  • Nonpyrogenic
  • Available in nonsterile or sterile
Specifications
Volume (Metric)250 μL
FilteredNon-filtered
SterilityNon-sterile
ColorClear
Certifications/ComplianceANSI
Clean ClaimsDNase, Rnase Free
For Use With (Application)Non-pyrogenic, used for stringent specifications under strict process controls
FeaturesNon-pyrogenic
FormatRack
Tip StyleStandard
PackagingSBS Rack
Quantity50 Racks/Cs.
For Use With (Equipment)Agilent/Velocity VPrep 8LT, Agilent/Velocity VPrep 96LT, Agilent Bravo 96LT