200 μL Universal Pipetter Tips: 200 μL, Wide Bore
Actual product may vary
Axygen
200 μL Universal Pipetter Tips: 200 μL, Wide Bore
Designed to fit a wide variety of single and multi-channel pipettes
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSterility
FIS14-222-730Sterile
FIS14-222-726Non-sterile
Catalog number FIS14-222-730
Price (SGD)
148.00
Pack
Sterility:
Sterile
Price (SGD)
148.00
Pack
Axygen™ Universal Tips are designed to fit a wide variety of single and multi-channel pipettors. These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free. They are available in Maxymum Recovery, beveled, nonsterile, sterile, non-filtered, and other features for specialized applications. Flexible walls and a series of internal sealing rings ensure a secure fit with less force required to load and eject.
(This is the suggested replacement for T-205-WB-C-L)
  • These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free.
  • Nonpyrogenic
  • Wide-Bore
Specifications
Volume (Metric)1 to 200 μL
SterilitySterile
ColorClear
AutoclavableAutoclavable
Clean ClaimsDNase, RNase, and Pyrogen free
FormatRack
Tip StyleGenomic/Wide Orifice, Universal
For Use With (Equipment)Universal Fit