Search Fisher Scientific
| Catalog Number | Sterility | Capacity (Metric) | Includes |
|---|---|---|---|
| FIS14-222-562 | Non-sterile | 10 mL | Tubes with Caps |
| FIS14-222-650 | Non-sterile | 5 mL | Tubes with caps |
| FIS14-222-563 | Sterile | 10 mL | Tubes with caps |
| FIS14-222-651 | Sterile | 5 mL | Tubes with caps |
For cap, see Cat. No. SC-B-510.