Multi Well High Profile Reagent Reservoirs
Actual product may vary
Multi Well High Profile Reagent Reservoirs
Manufactured with virgin polypropylene for excellent chemical resistance
Have Questions?
Catalog NumberCapacity (Metric)SterilityChannelsPackaging
FIS14-222-40284 mLNon-sterile1225/Cs.
Catalog number FIS14-222-402
Price (SGD)
831.00
Case
Capacity (Metric):
84 mL
Sterility:
Non-sterile
Channels:
12
Packaging:
25/Cs.
Price (SGD)
831.00
Case
Corning offers a complete line of Axygen single-well and multiple-well reagent reservoirs which are available in multiple formats for automated and manual use. All reservoirs are designed to comply with standard ANSI/SLAS footprint dimensions for microplates and are compatible with most automation workstations. Reagent reservoirs are manufactured with polypropylene for excellent chemical resistance.
  • Durable and cost effective
  • Various formats for multiple applications
Specifications
Certifications/ComplianceANSI/SLAS, RNase/DNase-free, non-pyrogenic
ColorClear
Capacity (Metric)84 mL
MaterialPolypropylene
SterilityNon-sterile
TypeReagent Reservoir
No. per Case25/Cs.
No. per Pack25/Pk.
Packaging25/Cs.
Volume (Metric) Well22 mL
Channels12