Caps for PCR Tubes
Actual product may vary
Axygen
Caps for PCR Tubes
8-Strip domed or flat strip caps for Axygen PCR tubes
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorClosure TypeProduct Type
FIS14-222-265ClearDomed Cap8-Strip Domed PCR Tube Cap
FKS12609535MaxyClear, w/writing surfaceFlat Cap8-Strip Flat PCR Tube Cap with Writing Surface
Catalog number FIS14-222-265
Price (SGD)
59.00
Pack
Color:
Clear
Closure Type:
Domed Cap
Product Type:
8-Strip Domed PCR Tube Cap
Price (SGD)
59.00
Pack
Axygen polypropylene, thin wall PCR tubes with and without caps are designed to fit standard 96-well blocks and are available in an assortment of colors. The consistent and ultra-thin wall allows precise thermal transfer. An even insertion cap ensures a tight fit to reduce evaporation.
Specifications
Certifications/ComplianceRNase, DNase, pyrogen-free
Closure ColorClear
ColorClear
Volume (Metric)0.2 mL
Quantity125/Pk.
Closure MaterialPolypropylene
For Use With (Equipment)8-strips PCR tubes and 96-well PCR microplates
SterilityNon-sterile
MaterialPolypropylene
Closure TypeDomed Cap
FormatStrip
Product Type8-Strip Domed PCR Tube Cap
Unit SizePack