12-Strip PCR Tubes
Actual product may vary
Axygen
12-Strip PCR Tubes
Manufactured from 99.9% pure virgin polypropylene
Have Questions?
Catalog NumberProduct Type
FIS14-222-259Thin Wall 12-Strip PCR Tube and Flat Cap
Catalog number FIS14-222-259
Price (SGD)
3,144.00
Case
Product Type:
Thin Wall 12-Strip PCR Tube and Flat Cap
Price (SGD)
3,144.00
Case
Axygen™ 0.2 mL thin-wall 12 strip PCR tubes, available in an assortment of colors are designed to fit a wide variety of thermal cyclers. Ultra-thin and consistent wall thickness allows precise thermal transfer, even insertion cap ensures a tight fit to prevent evaporation.
  • RNase-/DNase-free
  • Non-pyrogenic
  • Non-sterile
  • Manufactured with 99.9% pure virgin polypropylene
Specifications
Certifications/ComplianceRNase, DNase-free
ColorClear
Volume (Metric)0.2 mL
Quantity80/Pk.
Closure MaterialPolypropylene
For Use With (Equipment)Thermal Cyclers
SterilityNon-sterile
MaterialPolypropylene
Max. Force4000 x g
Closure TypeFlat Cap
FormatStrip
Product TypeThin Wall 12-Strip PCR Tube and Flat Cap
Unit SizeCase