Specifications
Product TypeDrum Thief
MaterialSoda Lime Glass
AutoclavableNot Autoclavable
Capacity (English)5.07 oz.
Capacity (Metric)150 mL
DisposableSingle-use
For Use With (Application)Ideal for quick and easy sample retrieval
Diameter (English) Outer0.7 in.
Diameter (Metric) Outer1.8 cm
Length (English)43 in.
Length (Metric)109.2 cm
Unit SizeCase