Wheaton™ Fritted Glass Support Base for use with Wheaton 47mm Filtration Assembly
Actual product may vary
DWK Life Sciences
Wheaton™ Fritted Glass Support Base for use with Wheaton 47mm Filtration Assembly
Replacement Glass Support Base with 40/35 [ST] Joint Connection for use with DWK Life Sciences Wheaton 47mm Glass Filter Holder Assembly (419380).
Have Questions?
Catalog NumberFor Use With (Equipment)
FIS14-209-126Filtration Assembly (47 mm)
Catalog number FIS14-209-126
Price (SGD)
-
For Use With (Equipment):
Filtration Assembly (47 mm)
  • Fritted glass support base for Wheaton 47mm Vacuum Filter Holder Assemblies
  • Manufactured from Wheaton 33 low extractable borosilicate glass which conforms to USP Type I and ASTM E-438 Type I, Class A requirements.
  • Coarse porosity (40 to 60μm)
Specifications
Certifications/ComplianceUSP Type I, ASTM E 438 Type I, Class A
For Use With (Equipment)Filtration Assembly (47 mm)
MaterialGlass
No. per Case1/Cs.
TypeFritted Support Base
Unit SizeCase