Bytac™ Surface Protector
Actual product may vary
Saint-Gobain
Bytac™ Surface Protector
Put the protection of FEP fluoropolymer on any surface with Bytac™ Surface Protectors.
Have Questions?
Catalog NumberDimensions (L x W)Material
FIS14-206-4415 ft. x 25 in.FEP, Vinyl
Catalog number FIS14-206-44
Price (SGD)
-
Dimensions (L x W):
15 ft. x 25 in.
Material:
FEP, Vinyl
Put the protection of FEP fluoropolymer on any surface
  • Made of an overlay of FEP film bonded to a support backing of vinyl or aluminum (backing is coated with pressure-sensitive acrylic adhesive)
  • Protect benchtops, fume hoods, temporary tables, walls/ceilings, splash screens, walls of spray booths, and other lab surfaces
  • Help protect surfaces against absorption of toxic or radioactive chemicals
  • Corrosive chemicals, paint, oil, grease, and sticky substances wipe off easily with damp sponge
  • Cut easily with scissors
  • Install easily—simply peel off protective paper from adhesive backing and apply to surface
Specifications
Product TypeBench Protector
MaterialFEP, Vinyl
Length (English)15 ft.
Length (Metric)4.6 m
Width (English)25 in.
Width (Metric)640 mm
Dimensions (L x W)15 ft. x 25 in.
For Use With (Application)Protects benchtops, fume hoods, temporary tables, walls/ceilings, splash screens, walls of spray booths, and other lab surfaces
Quantity1 Ea.
Unit SizeEach