Check Order Status
Natural Rubber Latex Tubing on 50ft. (15.2m) Reel
Actual product may vary
Natural Rubber Latex Tubing on 50ft. (15.2m) Reel
Contains dry natural rubber
Have Questions?
Change view
Catalog Number
Diameter (English) Inner
Diameter (English) Outer
Thickness (English)
FIS14-178-2CC
1/4 in.
0.44 in.
0.0944 in.
FIS14-178-2B
0.188 in.
0.31 in.
0.062 in.
2 Options
Catalog number
FIS14-178-2CC
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Diameter (English) Inner:
1/4 in.
Diameter (English) Outer:
0.44 in.
Thickness (English):
0.0944 in.
Contains dry natural rubber
Made with dry natural rubber
Available in variety of sizes
Amber colored
Specifications
Certifications/Compliance
ZZ-T-831D Types I and III, U.S. Pharmacopeia Class VI
Autoclavable
Autoclavable
Diameter (English) Inner
1/4 in.
Diameter (Metric) Inner
6.4 mm
Material
Rubber
Diameter (English) Outer
0.44 in.
Diameter (Metric) Outer
11.1 mm
Overall Length (English)
50 ft.
Overall Length (Metric)
15.2 m
Thickness (English)
0.0944 in.
Thickness (Metric)
2.4 mm
Color
Amber
Product Type
Tubing