Pressure and Vacuum Rubber Tubing
Pressure and Vacuum Rubber Tubing
Actual product may vary
Pressure and Vacuum Rubber Tubing
Pressure and Vacuum Rubber Tubing
Pressure and Vacuum Rubber Tubing
Fisher Catalogue
Pressure and Vacuum Rubber Tubing
For pressure and vacuum work
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (English) InnerDiameter (English) OuterOverall Length (English)Color
FIS14-175E3/8 in.7/8 in.12 ft.Black
FIS14-150-2K5/16 in.11/16 in.50 ft.Black
FIS14-173D3/8 in.1 in.12 ft.Red
FIS14-150-2J1/4 in.0.62 in.50 ft.Black
FIS14-175D5/16 in.11/16 in.12 ft.Black
Catalog number FIS14-175E
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Diameter (English) Inner:
3/8 in.
Diameter (English) Outer:
7/8 in.
Overall Length (English):
12 ft.
Color:
Black
For pressure and vacuum work
  • Made with dry natural rubber
  • Available in variety of sizes
  • Offered in Black and Red
Specifications
Diameter (English) Inner3/8 in.
Diameter (Metric) Inner9.5 mm
MaterialRubber
Diameter (English) Outer7/8 in.
Diameter (Metric) Outer22.2 mm
Overall Length (English)12 ft.
Overall Length (Metric)3.7 m
Thickness (English)0.25 in.
Thickness (Metric)6.4 mm
For Use With (Application)Pressure and vacuum work
ColorBlack
Product TypePressure and Vacuum Tubing
Unit SizePK