Tygon S3™ E-3603 Flexible Tubings
Actual product may vary
Saint-Gobain
Tygon S3™ E-3603 Flexible Tubings
Formulated for resistance to flex-fatigue and abrasion.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (English) InnerDiameter (English) OuterThickness (English)
FIS14-171-2255/16 in.9/16 in.1/8 in.
FIS14-171-1291/16 in.1/8 in.1/32 in.
FIS14-171-1303/32 in.5/32 in.1/32 in.
FIS14-171-2111/8 in.3/16 in.1/32 in.
FIS14-171-1031/8 in.1/4 in.1/16 in.
FIS14-171-2153/16 in.1/4 in.1/32 in.
FIS14-171-2163/16 in.5/16 in.1/16 in.
FIS14-171-1313/16 in.7/16 in.1/8 in.
FIS14-171-1041/4 in.3/8 in.1/16 in.
FIS14-171-1321/4 in.7/16 in.3/32 in.
FIS14-171-2211/4 in.1/2 in.1/8 in.
FIS14-171-2235/16 in.7/16 in.1/16 in.
FIS14-171-2245/16 in.1/2 in.3/32 in.
FIS14-171-2273/8 in.1/2 in.1/16 in.
FIS14-171-2283/8 in.9/16 in.3/32 in.
FIS14-171-1333/8 in.5/8 in.1/8 in.
FIS14-171-2351/2 in.11/16 in.3/32 in.
FIS14-171-2361/2 in.3/4 in.1/8 in.
FIS14-171-2425/8 in.7/8 in.1/8 in.
FIS14-171-2453/4 in.1 in.1/8 in.
FIS14-171-2521 in.1 1/4 in.3/8 in.
Catalog number FIS14-171-225
Price (SGD)
-
Diameter (English) Inner:
5/16 in.
Diameter (English) Outer:
9/16 in.
Thickness (English):
1/8 in.
Formulated for resistance to flex-fatigue and abrasion.
  • Bio-based formulation and contain no BPA or phthalates
  • Provide high chemical resistance
  • Non-oxidizing and non-contaminating
  • Smooth, polished inner wall helps prevent buildup to facilitate cleaning
  • Coils are marked at 30.4cm (1 ft.) intervals for easy measuring
  • Slips easily over fittings and grips securely for simple lab set-ups
  • Extra-heavy walls withstand full vacuum at room temperature (759mm [29.9 in.] of mercury at 23°C [73°F] and up to 686mm [27 in.] of mercury at 60°C [140°F])
  • Sterilized by conventional autoclave methods (steam 30 min. at 15psi, 121°C) and Ethylene Oxide
Applications:
  • Instrumentation connection, vent, drain and other general laboratory applications
  • Food and beverage
  • Biopharmaceutical
  • Analytical instruments
  • Peristaltic and vacuum pumps
  • Condensers
  • Incubators
  • Desiccators
  • Gas lines
  • Drain lines
Specifications
Diameter (English) Inner5/16 in.
Diameter (English) Outer9/16 in.
Overall Length (English)50 ft.
Overall Length (Metric)15 m
Thickness (English)1/8 in.