Tygon™ E-3603 Tubing
Actual product may vary
Tygon™ E-3603 Tubing
Tygon™ E-3603 Tubing
Saint-Gobain
Tygon™ E-3603 Tubing
Tygon™ E-3603 Tubing for laboratory and vacuum applications.
Have Questions?
Catalog NumberDiameter (English) InnerDiameter (English) OuterMaximum PressureThickness (English)
FIS14-171-2221/4 in.0.625 in.53 psi (at 73°F)3/16 in.
Catalog number FIS14-171-222
Price (SGD)
-
Diameter (English) Inner:
1/4 in.
Diameter (English) Outer:
0.625 in.
Maximum Pressure:
53 psi (at 73°F)
Thickness (English):
3/16 in.
Tygon™ E-3603 tubing from Saint-Gobain is crystal clear and flexible, handling virtually all organic chemicals found in the modern laboratory. It is non-oxidizing and non-contaminating, making it an ideal choice for a variety of applications. Tygon™ E-3603 has high tolerances and lot-to-lot consistency, which will yield reproducible results in your laboratory. It is non-toxic and non-contaminating, and less permeable than rubber tubing. The glassy-smooth inner bore helps prevent buildup to facilitate cleaning.
  • Lot-to-lot consistency for reproducible results
  • Non-toxic and non-contaminating
  • Smooth, polished inner wall
  • Slips easily over fittings and grips securely for simple lab set-ups
  • Contains no BPA or phthalates
  • Standard sizes available to hold full vacuum at room temperature
Specifications
Diameter (English) Inner1/4 in.
Diameter (Metric) Inner6.4 mm
MaterialTygon S3 E-3603
Diameter (English) Outer0.625 in.
Diameter (Metric) Outer15.9 mm
Overall Length (English)50 ft.
Overall Length (Metric)15 m
Thickness (English)3/16 in.
Thickness (Metric)4.8 mm
ColorClear
Flexibility13 mm bend radius
Maximum Pressure53 psi (at 73°F)
Product TypeTubing
Unit SizePack