Two-Hole Rubber Stoppers
Two-Hole Rubber Stoppers
Actual product may vary
Two-Hole Rubber Stoppers
Have Questions?
Catalog NumberStopper Number
FIS14-140H6
Catalog number FIS14-140H
Price (SGD)
82.00
PK
Add to cart
Stopper Number:
6
Price (SGD)
82.00
PK
Add to cart
  • Hole size: 3 to 5mm
Specifications
Diameter (Metric) Hole0.5cm
Diameter (Metric) Inner5mm
For Use With (Application)General purpose use
Length (Metric)2.5 cm
Size6
Total Positions2
ColorBlack
Diameter (Metric) Bottom2.6 cm
Diameter (Metric) Top3.2 cm
MaterialNatural Rubber
Product TypeTwo Hole Taper Stopper
Stopper Number6
Unit SizePK