One-Hole Rubber Stoppers
One-Hole Rubber Stoppers
Actual product may vary
One-Hole Rubber Stoppers
Available in Size No. 00 to 15 Hole sizes 3 to 5mm
Have Questions?
Catalog NumberStopper Number
FIS14-135J6
Catalog number FIS14-135J
Price (SGD)
84.00
PK
Add to cart
Stopper Number:
6
Price (SGD)
84.00
PK
Add to cart
One hole rubber stopper
  • Available in Size No. 00 to 15
  • Hole size: 3 to 5mm
Specifications
Diameter (Metric) Hole0.5cm
Diameter (Metric) Inner5mm
For Use With (Application)Inexpensive black natural rubber/SBR blend for general purpose uses
Length (Metric)0.25 cm
Size6
Total Positions1
ColorBlack
Diameter (Metric) Bottom2.6 cm
Diameter (Metric) Top3.2 cm
MaterialNatural Rubber
Product TypeOne Hole Taper Stopper
Stopper Number6
Unit SizePK