Check Order Status
Solid Rubber Stoppers
Actual product may vary
Solid Rubber Stoppers
Available in a variety of sizes ranging from 000 through 16
Have Questions?
Change view
Catalog Number
Size
Stopper Number
Length (Metric)
FIS14-130U
13
13
2.5 cm
FIS14-130H
5.5
5.5
2.5 cm
FIS14-130A
00
0
2.5 cm
FIS14-130Q
10.5
10.5
2.5 cm
FIS14-130L
7
7
2.5 cm
5 Options
Catalog number
FIS14-130U
Price (SGD)
80.00
PK
Add to cart
Size:
13
Stopper Number:
13
Length (Metric):
2.5 cm
Price (SGD)
80.00
PK
Add to cart
Available in a variety of sizes ranging from 000 through 16
Solid rubber construction
Specifications
For Use With (Application)
General purpose use
Length (Metric)
2.5 cm
Size
13
Color
Black
Diameter (Metric) Bottom
5.8 cm
Diameter (Metric) Top
6.8 cm
Material
Natural Rubber
Product Type
Solid Stopper
Stopper Number
13