Check Order Status
Solid Rubber Stoppers
Actual product may vary
Solid Rubber Stoppers
Available in a variety of sizes ranging from 000 through 16
Have Questions?
Catalog Number
Stopper Number
FIS14-130J
6
Catalog number
FIS14-130J
Price (SGD)
76.00
PK
Add to cart
Stopper Number:
6
Price (SGD)
76.00
PK
Add to cart
Available in a variety of sizes ranging from 000 through 16
Solid rubber construction
Specifications
For Use With (Application)
General purpose use
Length (Metric)
2.5 cm
Size
6
Color
Black
Diameter (Metric) Bottom
2.6 cm
Diameter (Metric) Top
3.2 cm
Material
Natural Rubber
Product Type
Solid Stopper
Stopper Number
6