▸
Biochemical Reagents
▸
Staining Reagents
▸
Stains and Dyes
▸
Bromothymol Blue Indicator, 0.04% (w/v), Ricca Chemical
Element Search
Structure Search
Bromothymol Blue Indicator, 0.04% (w/v), Ricca Chemical
For Glassware Residue Detection
Have Questions?
Catalog Number
Quantity
Packaging
FIS1375-16
500 mL
Natural Poly Bottle
Catalog number
FIS1375-16
Price (SGD)
-
Quantity:
500 mL
Packaging:
Natural Poly Bottle
Request bulk or custom format
Chemical Identifiers
CAS
34722-90-2, 7732-18-5
Specifications
Chemical Name or Material
Bromothymol Blue Indicator
CAS
34722-90-2
CAS Min %
98.0
CAS Max %
100.0
CAS
7732-18-5
View more