Storage Boxes
Actual product may vary
Storage Boxes
Storage Boxes
Storage Boxes
Storage Boxes
Thermo Scientific
Storage Boxes
Store a variety of vials and tubes with these fiberboard storage boxes, for use in ultra-low temperature freezers.
Have Questions?
Catalog NumberHoldsDimensions (L x W x H)
FIS13-994-07081 Cryo Tubes5 x 5 x 2 in.
Catalog number FIS13-994-070
Price (SGD)
34.00
Each
Holds:
81 Cryo Tubes
Dimensions (L x W x H):
5 x 5 x 2 in.
Price (SGD)
34.00
Each
Store a variety of vials and tubes with Thermo Scientific™ Storage Boxes, for use in ultra-low temperature freezers.
  • Sturdy, water-repellent boxes with covers for sample storage
  • Available in 2 and 3 in. high (5.1 and 7.6 cm) with 5.25 x 5.25 in. (13.3 x 13.3 cm) square bases
  • Box dividers included excluding 5954 and 5956
Specifications
ColorWhite
MaterialFiberboard
ShapeSquare
TypeCryo Storage Box
For Use With (Equipment)Thermo Scientific Ultra-Low Temperature Freezers
Holds81 Cryo Tubes
IncludesDividers
Dimensions (L x W x H)5 x 5 x 2 in.
Length (English)5 in.
Length (Metric)12.7 cm
Width (English)5 in.
Width (Metric)12.7 cm
Height (English)2 in.
Height (Metric)5 cm
Weight (Metric)0.99 kg
Unit SizeEach