Thermo Scientific
Temperature Chart Recorder Paper
Use Thermo Scientific™ Temperature Chart Recorder Paper in seven-day circular temperature chart recorders.
Have Questions?
Catalog NumberTemperature Range
FIS13-990-16A-40°C to +25°C
Catalog number FIS13-990-16A
Price (SGD)
575.00
Pack
Temperature Range:
-40°C to +25°C
Price (SGD)
575.00
Pack
Use Thermo Scientific™ Temperature Chart Recorder Paper in seven-day circular temperature chart recorders.
  • 6 in. (15.2cm) diameter
  • For use in freestanding and panel-mounted chart recorders
Specifications
TypeChart Recorder Paper
Temperature Range-40°C to +25°C
For Use With (Equipment)Free-Standing and Panel-Mounted Chart Recorders
Monitoring OptionsRecorder
Static Holding Time7 Days
Diameter (English)6 in.
Diameter (Metric)15.2 cm
Unit SizePack