BIO-CERT™ Pipette Tips, 0.5 to 20 μL
Actual product may vary
BrandTech
BIO-CERT™ Pipette Tips, 0.5 to 20 μL
DNA-, RNase-, ATP- and endotoxin-free
Have Questions?
Catalog NumberTip StyleFormatPackaging
FIS13-889-178NarrowBoxTipRack
Catalog number FIS13-889-178
Price (SGD)
126.00
Each
Tip Style:
Narrow
Format:
Box
Packaging:
TipRack
Price (SGD)
126.00
Each
  • High-purity polypropylene is oleamide- and diHEMDA-free
  • Independently certified sterile and free from biological contaminants
  • Fits pipettes using ultra-micro/'crystal' tips
  • Slender design
  • Graduation at 2 and 10 μL for quick volume check
  • Available in either standard or ultra low retention types
  • Available in convenient, ecological rack-and-stack formats
  • Tips are colorless and placed into gray-colored tip tray
Ultra low retention tips
  • Produced through special, patented physicochemical process
  • Homogeneous, defect-free surface for low surface tension and low sample retention
  • Reduce sample loss and provide substantially higher reproducibility when working with critical media
  • For biological samples that contain proteins, nucleic acids, detergents and other difficult liquids that can cause sample loss
  • High chemical resistance
  • Tips can be autoclaved at 121°C (2 bar) without damaging low retention properties
Specifications
DescriptionBIO-CERT™ Liquid Handling Quality
Volume (Metric)0.5 to 20 μL
FilteredNon-filtered
SterilitySterile
ColorColorless
Rack ColorGray
AutoclavableAutoclavable
Length (English)1.81 in.
Length (Metric)4.6 cm
Clean ClaimsDNase, RNase, ATP, and Endotoxin Free
For Use With (Application)High translucency, easy ejection and low waste, quick volume check
Sterilization MethodBeta Radiation
FormatBox
Tip StyleNarrow
PackagingTipRack
Product TypePipette Tips
DisposableLimited-use
For Use With (Equipment)Universal Fit Ultramicro
GraduatedYes
Product LineBio-Cert
MaterialPolypropylene
Graduations2, 10 μL
Unit SizeEach