Pipette Tips, 0.5 to 20 μL
Actual product may vary
BrandTech
Pipette Tips, 0.5 to 20 μL
Manufactured from high-purity, oleamide and diHEMDA-free polypropylene. Pipet Tips, 0.5 to 20μL allow pipetting in microtubes and microplates without touching the walls. Gradations marked at 2μL and 10μL provide quick volume assessment.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberPackagingQuantityFilteredTip Style
FIS13-889-172TipBox480/Pk.Non-filteredNarrow
FKS15114897Bulk10,000/Pk.Non-filteredNarrow
Catalog number FIS13-889-172
Price (SGD)
62.00
Pack
Packaging:
TipBox
Quantity:
480/Pk.
Filtered:
Non-filtered
Tip Style:
Narrow
Price (SGD)
62.00
Pack
  • Nonsterile , high-purity polypropylene is free from oleamide and diHEMDA.
  • Gradations marked at 2 μL and 10 μL provide quick volume assessment.
  • Slender design allows pipetting in microtubes and microplates without touching the walls.
  • Available in either standard or ultra low retention types.
  • Available either in bulk packages or in convenient rack-and-stack formats.
Ultra low retention tips
  • Produced through special, patented physicochemical process.
  • Homogeneous, defect-free surface thus produced has low surface tension.
  • Reduce sample loss and provide substantially higher reproducibility when working with critical media.
  • For biological samples that contain detergents such as Triton™ X-100, SDS, Tween, etc.
  • High chemical resistance.
  • Tips can be autoclaved at 121°C (2 bar) without damaging material properties.
Specifications
DescriptionBIO-CERT™ Liquid Handling Quality
Volume (Metric)0.5 to 20 μL
FilteredNon-filtered
SterilityNon-sterile
ColorColorless
Rack ColorGray
AutoclavableAutoclavable
Length (English)1.8 in.
Length (Metric)46 mm
Clean ClaimsDNase, RNase, ATP, and Endotoxin Free
For Use With (Application)Reverse pipetting, plate washing, working with multichannel pipettes
FormatBox
Tip StyleNarrow
PackagingTipBox
Quantity480/Pk.
Product TypePipette Tips
DisposableLimited-use
For Use With (Equipment)Universal Fit Ultramicro
GraduatedYes
MaterialPolypropylene
Fit for PurposePCR Ready
Graduations2, 10 μL
Unit SizePack