UV-Cuvettes - CERTIFIED LIFE SCIENCE QUALITY
UV-Cuvettes - CERTIFIED LIFE SCIENCE QUALITY
Actual product may vary
UV-Cuvettes - CERTIFIED LIFE SCIENCE QUALITY
UV-Cuvettes - CERTIFIED LIFE SCIENCE QUALITY
BrandTech
UV-Cuvettes - CERTIFIED LIFE SCIENCE QUALITY
BRAND™ disposable UV-cuvettes are a cost-effective, durable alternative to fragile quartz cuvettes in numerous applications
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Height (Metric)QuantityWidth (Metric)
FIS13-878-12170 to 850 μL45 mm100 Pack12.5 mm
FIS03-840-30070 to 850 μL45 mm100 Pack, Individually Wrapped12.5 mm
FIS13-688-731.5 to 3 mL45 mm500 Pack12.5 mm
FIS13-688-741.5 to 3 mL45 mm100 Pack12.5 mm
FKS105665811.5 to 3 mL45 mm100 Pack12.5 mm
FKS1029027270 to 850 μL45 mm100 Pack12.5 mm
FIS13-878-12370 to 550 μL45 mm100 Pack12.5 mm
FIS13-878-12470 to 550 μL45 mm500 Pack12.5 mm
FIS14-377-0092.5 to 4.5 mL45 mm100 Pack12.5 mm
FKS100467312.5 to 4.5 mL45 mm100 Pack12.5 mm
Catalog number FIS13-878-121
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
70 to 850 μL
Height (Metric):
45 mm
Quantity:
100 Pack
Width (Metric):
12.5 mm
BrandTech™ BRAND™ UV-Cuvette Disposable Cuvets provide transparency from 230 to 900nm and eliminate cleaning and risk of cross-contamination. Ideal for DNA, RNA and protein analysis. Compatible with with most polar organic solvents, acids, and bases.
CERTIFIED LIFE SCIENCE QUALITY cuvettes from BRAND™ are manufactured from virgin raw materials under controlled room conditions, with intensive in-process controls, visual inspections, batch management, and final product inspection to ensure proven functionality.
  • Advanced cuvette design includes recessed windows to protect against scratching in the optical pathway
  • Disposable - cleaning and cross-contamination risks are eliminated
  • Ideal for DNA, RNA, and protein analysis
  • Chemical resistant - can be used with most polar organic solvents, acids, and bases
  • Also compatible with acetone, butanone, DMF, and concentrated hydrochloric acid
  • Grouped by manufacturing mold cavity to ensure lowest variation in extinction coefficient
  • Fits most spectrophotometers without adapters
Ultramicro
  • Available with 8.5 and 15 mm window heights (Z-dimension)
  • May be used with sample volumes as small as 70 μL
  • Can be fitted with plug seal caps (sold separately) for sample storage
  • Individually wrapped UV-Cuvettes are DNA-, DNase-, and RNase-free
Semimicro
  • For sample volumes between 1.5 and 3.0 mL
Macro
  • For sample volumes between 2.5 and 4.5 mL
Specifications
SterilityNon-sterile
TypeCuvette
ColorClear
DisposableSingle-use
Capacity (Metric)70 to 850 μL
MaterialPlastic
IncludesCuvette Only
Quantity100 Pack
Path Length10 mm
Wavelength Range230 to 900 nm
Height (Metric)45 mm
For Use With (Equipment)Spectrophotometers and photometers
Width (Metric)12.5 mm
For Use With (Application)DNA, RNA, Protein Analysis, Molecular Biology
Lightpath10 mm
ShapeRectangle
Manufacturing QualityCERTIFIED LIFE SCIENCE QUALITY
Unit SizePK