Stomacher™ 80 Strainer Bags
Actual product may vary
Seward
Stomacher™ 80 Strainer Bags
The Stomacher™ Bag range has been designed to fulfil all applications from sample blending, straining and storage, including bags designed for extensive blending and small samples for life science applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
FIS13-874-756Filter, Closure, Strainer Bag
FIS13-874-754Filter/Strainer Bag
Catalog number FIS13-874-756
Price (SGD)
876.00
Each
Description:
Filter, Closure, Strainer Bag
Price (SGD)
876.00
Each
The Stomacher™ Bag range has been designed to fulfil all applications from sample blending, straining and storage, including bags designed for extensive blending and small samples for life science applications.
  • Seward Stomacher bag range is made from high-quality food-grade virgin LDPE granule extruded to 65μm film thickness
  • Heavy-duty bags also available for large volume sampling 80μm film thickness; Sterile (gamma-irradiated) polyethylene bags; Closure bags feature wire closure strip across bag top for short term storage.
  • Strainer bags have inner plastic mesh to make it easy to pipette from samples that produce a large amount of debris
  • Single-chamber bags for use in Lab Blenders; internal sterility assured with proprietary tear-off sterile barrier top on sachets
  • Highly transparent and puncture-resistant virgin polyethylene extruded tube material
  • Simply tear the perforated end along the top of the sachet, remove the bag and open using Seward bag opener, insert the material to be analyzed and processed
  • Bags available for sample size 25μl - 4500ml
Specifications
DescriptionFilter, Closure, Strainer Bag
AutoclavableNot Autoclavable
Closure TypeFlat Wire
ColorClear
DisposableSingle-use
For Use With (Application)To prepare, store and transport samples in microbiology sample preparation
For Use With (Equipment)Stomacher 80 type lab blenders
MaterialLDPE
SterilitySterile
No. per Pack200
Quantity200/Pk.
Sterilization MethodIrradiated
Dimensions (L x W)15.5 x 10.5cm (6 x 4 in.)
Product TypeStrainer Bag
Capacity (Metric)5 to 80 mL
Length (Metric)15.5 mm
Unit SizeEach