Aluminum Baskets with Tilt-Top Lids
Actual product may vary
Fisherbrand
Aluminum Baskets with Tilt-Top Lids
Fisherbrand Aluminum Baskets with Tilt-Top Lids are ideal for rinsing or storing tubes and confining small lab animals
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDimensions (L x W x H)
FIS13-807-1812.87 x 9 x 7 in.
FIS13-807-116 x 6 x 6 in.
Catalog number FIS13-807-18
Price (SGD)
159.00
Each
Dimensions (L x W x H):
12.87 x 9 x 7 in.
Price (SGD)
159.00
Each
Fisherbrand Aluminum Baskets with Tilt-Top Lids are ideal for rinsing or storing tubes and confining small lab animals
  • Fisherbrand™ Aluminum Baskets with Tilt-Top Lids are ideal for rinsing or storing tubes and confining small lab animals
  • Have many applications and various shapes and sizes are available
  • Diamond Flow-Through pattern allows for maximum penetration of sterilizer solution
  • Reinforced top, bottom and seams welded to frame plate
  • Rust and corrosion resistive
  • Baskets tilt at an optimum angle for easy dispening of test tubes and other labware and instruments
  • Tilt-cover doubles as angle supports
  • Also used to store animals
Specifications
Closure IncludedYes
AutoclavableNot Autoclavable
MaterialAluminum
ShapeRectangular
SterilitySterile
Dimensions (L x W x H)12.87 x 9 x 7 in.
Length (Metric)32 cm
Length (English)12.87 in.
Width (English)9 in.
Height (English)7 in.
Product TypeAluminum Basket
Height (Metric)18 cm
Width (Metric)23 cm
Unit SizeEach