Specifications
Capacity4 oz., 125 mL
Capacity (Metric)125 mL
Capacity (English)4 oz.
ColorClear
Diameter (English)2 in.
For Use With (Application)Liquid and Dry Storage and Packaging
IncludesCap in Bag
MaterialSoda Lime Glass
PackagingConvenience Pk. (caps attached)
Closure ColorBlack
ClarityClear
Certifications/ComplianceManufactured from Soda Lime Glass Conforming to USP Type III and ASTM E438, Type II Requirements
MouthWide
ShapeRound
Height (Metric)84 mm
Product TypeJar
Height (English)3.3 in.
Closure LinerPulp/Vinyl
Closure MaterialPhenolic
Closure Size48-400 mm
Diameter (Metric)52 mm
Closure TypeCap
Unit SizeCase