Biohazard Disposal Bags with or without Warning Label
Actual product may vary
Bel-Art
Biohazard Disposal Bags with or without Warning Label
Economical, safe disposal of used pipettes, petri dishes, multi-well cell culture plates, cell culture flasks, culture plates, membrane filters, and other biohazard wastes
Have Questions?
Catalog NumberDimensions (L x W)Color
FIS13-717-28724 x 12 in. (61 x 30 cm)Translucent
Catalog number FIS13-717-287
Price (SGD)
202.00
Pack
Dimensions (L x W):
24 x 12 in. (61 x 30 cm)
Color:
Translucent
Price (SGD)
202.00
Pack
Economical, safe disposal of used pipettes, petri dishes, multi-well cell culture plates, cell culture flasks, culture plates, membrane filters, and other biohazard wastes
  • These transparent, polypropylene plastic biohazard bags are 0.03 mm (1.5 mil) thick
  • High temperature autoclavable at a maximum of 135°C (275°F)
  • May also be used as conventional trash can liners
Specifications
Capacity (Metric)15 L
Certifications/ComplianceASTM D1709-98
Closure TypeZip Tie
DesignNon-printed without Sterilization Patch
Capacity (English)3 gal.
ColorTranslucent
Dimensions (L x W)24 x 12 in. (61 x 30 cm)
DisposableSingle-use
Length (Metric)61 cm
SterilityNon-sterile
Width (Metric)30 cm
AutoclavableAutoclavable
For Use With (Application)Disposal of used pipettes, petri dishes, culture plates, membrane filters and more
Width (English)12 in.
Length (English)24 in.
Thickness1.5 mil (0.03 mm)
MaterialPolypropylene
Product TypeBiohazard Bag
Unit SizePack