Protein LoBind™ Tubes
Actual product may vary
Protein LoBind™ Tubes
Protein LoBind™ Tubes
Protein LoBind™ Tubes
Eppendorf
Protein LoBind™ Tubes
Engineered to minimize sample binding and elevate recovery, Eppendorf™ Protein LoBind™ Tubes safeguard your valuable proteins during processing. With cutting-edge LoBind technology, ensure optimal yields for your Western blotting, mass spectrometry, and protein purification applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FIS13-698-7930.5 mL
FIS13-698-7941.5 mL
FIS13-698-7952 mL
FISE9250000901.5 mL
Catalog number FIS13-698-793
Price (SGD)
127.00
Pack
Capacity (Metric):
0.5 mL
Price (SGD)
127.00
Pack
  • Surface-coating-free, including substances like silicone, to reduce the risk of sample interference.
  • Lot-certified PCR clean purity grade guarantees freedom from human DNA, DNase, RNase, and PCR inhibitors.
  • Available in tube, microplate, and deep-well plate formats for convenient upscaling of experiments.
  • Snap, screw, and SnapTec™ cap options available.
Specifications
Certifications/ComplianceDNA, DNase, RNase, PCR Inhibitors free
Closure MaterialHDPE
ColorClear
Capacity (Metric)0.5 mL
MaterialPolypropylene
For Use WithProtein LoBind consumables are best suited for work in protein research, mass spectrometry, protein array sample preparation, and storage of peptide, antibody or virus samples
IncludesSafe-lock lid
No. per Pack100 Pk.
Packaging100 Tubes (2 Bags x 50 Tubes)
SterilityNon-sterile
RNase FreeYes
Dnase FreeYes
AutoclavableAutoclavable
ShapeCylindrical
Capacity (English)0.02 oz.
Clean ClaimsFree of surface coating (e.g. silicone), plasticizers, biocides or mold release agents
Bottom ShapeConical
Closure TypeSnap Cap
Product TypeMicrocentrifuge Tube (PCR Clean, Protein LoBind)
Max. RCF18,000 x g
Purity or Quality GradePCR Clean (Non-sterile)
Unit SizePack