Dispensette™ S Analog-adjustable Bottletop Dispensers
Actual product may vary
Dispensette™ S Analog-adjustable Bottletop Dispensers
Dispensette™ S Analog-adjustable Bottletop Dispensers
BrandTech
Dispensette™ S Analog-adjustable Bottletop Dispensers
The Dispensette™ S Analog Bottletop Dispensers feature a 'floating piston' design for low-force operation and no wearing parts. Optimized flow channels make dispensing and priming even easier-particularly in instruments with large volumes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)AccuracyIncludesIncrements
FIS13-689-0162.5 to 25 mL0.5% (125μL)Dispensette S bottle-top dispenser,a GL 45 threaded bottle adapter, DE-M marking, performance certificate, telescoping filling tube, recirculation valve with an optional recirculation tube, a mounting tool, and polypropylene adapters (GL 32-33, GL38, S40)0.5 mL
FIS13-689-0130.2 to 2 mL0.5% (10μL)Dispensette S bottle-top dispenser,a GL 45 threaded bottle adapter, DE-M marking, performance certificate, telescoping filling tube, recirculation valve with an optional recirculation tube, a mounting tool, and polypropylene adapters (GL 24-25, GL 28/S28, GL 32-33, GL38, S40)0.05 mL
Catalog number FIS13-689-016
Price (SGD)
2,884.00
Each
Volume (Metric):
2.5 to 25 mL
Accuracy:
0.5% (125μL)
Includes:
Dispensette S bottle-top dispenser,a GL 45 threaded bottle adapter, DE-M marking, performance certificate, telescoping filling tube, recirculation valve with an optional recirculation tube, a mounting tool, and polypropylene adapters (GL 32-33, GL38, S40)
Increments:
0.5 mL
Price (SGD)
2,884.00
Each
  • New discharge tube, with or without recirculation valve
  • New valve system - no additional sealing rings required, facilitates cleaning and preparations for autoclaving
  • Simple priming by optimized flow channels
  • Less dispensing force - especially with large volume instruments
  • Safe volume setting with internal scalloped track (analog-adjustable instruments)
  • Pivoted screw cap - does not disturb dispensing
  • Easy to calibrate and adjust in order to comply with GLP guidelines
  • Easy to dismantle for cleaning
  • Replaceable filling and discharge valve with safety bulb (locks when discharge tube is not assembled)
  • 360° rotatable valve block allows bottle label to face user
  • Telescoping filling tube adjusts easily to different size bottles
  • DE-M marking
Specifications
Accuracy0.5% (125μL)
Product TypeBottletop Dispenser
Angle90°
AutoclavableAutoclavable
Certifications/ComplianceISO 9001
ColorGray/Red
DensityUp to 2.2 g/cm3
DescriptionWith recirculation valve
Channels1
MaterialBorosilicate Glass, Ceramic, ETFE, PFA, PTFE
Volume (Metric)2.5 to 25 mL
DisplayAnalog
For Use With (Application)Dispensing aggressive reagents, including concentrated bases and acids such as H3PO4, H2SO4 (except e.g., HCl, HNO3 und HF), saline solutions, as well as many organic solvents
IncludesDispensette S bottle-top dispenser,a GL 45 threaded bottle adapter, DE-M marking, performance certificate, telescoping filling tube, recirculation valve with an optional recirculation tube, a mounting tool, and polypropylene adapters (GL 32-33, GL38, S40)
Increments0.5 mL
Precision0.1% (25 μL)
Temperature (English)59°F to 104°F
Temperature (Metric)15°C to 40°C
Vapor PressureUp to 600 mbar
Viscosity RatingUp to 500 mm 2/s
Warranty2 Year
Unit SizeEach