Dispensette™ S Digital Bottletop Dispensers
Dispensette™ S Digital Bottletop Dispensers
Actual product may vary
Dispensette™ S Digital Bottletop Dispensers
Dispensette™ S Digital Bottletop Dispensers
BrandTech
Dispensette™ S Digital Bottletop Dispensers
The Dispensette™ S Digital Bottletop Dispensers feature a 'floating piston' design for low-force operation and no wearing parts. Optimized flow channels make dispensing and priming even easier-particularly in instruments with large volumes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberIncludesVolume (Metric)IncrementsAccuracy
FIS13-689-004Dispensette S bottle-top dispenser,a GL 45 threaded bottle adapter, DE-M marking, performance certificate, telescoping filling tube, recirculation valve with an optional recirculation tube, a mounting tool, and polypropylene adapters (GL 32-33, GL38, S40)2.5 to 25 mL0.1 mL0.5% (125μL)
FIS13-689-005Dispensette S bottle-top dispenser,a GL 45 threaded bottle adapter, DE-M marking, performance certificate, telescoping filling tube, recirculation valve with an optional recirculation tube, a mounting tool, and polypropylene adapters (GL 32-33, GL38, S40)5 to 50 mL0.2 mL0.5% (250μL)
FIS13-689-010Dispensette S bottle-top dispenser,a GL 45 threaded bottle adapter, DE-M marking, performance certificate, telescoping filling tube, a mounting tool, and polypropylene adapters (GL 32-33, GL38, S40)2.5 to 25 mL0.1 mL0.5% (125μL)
Catalog number FIS13-689-004
Price (SGD)
3,282.00
EA
Add to cart
Includes:
Dispensette S bottle-top dispenser,a GL 45 threaded bottle adapter, DE-M marking, performance certificate, telescoping filling tube, recirculation valve with an optional recirculation tube, a mounting tool, and polypropylene adapters (GL 32-33, GL38, S40)
Volume (Metric):
2.5 to 25 mL
Increments:
0.1 mL
Accuracy:
0.5% (125μL)
Price (SGD)
3,282.00
EA
Add to cart
The Dispensette™ S Digital Bottletop Dispensers feature a floating piston design for low-force operation and no wearing parts. Optimized flow channels make dispensing and priming even easier-particularly in instruments with large volumes. Designed for g
  • New discharge tube, with or without recirculation valve
  • New valve system - no additional sealing rings required, facilitates cleaning and preparations for autoclaving
  • Simple priming by optimized flow channels
  • Less dispensing force - especially with large volume instruments
  • Easy Calibration technique for calibration adjustment in seconds without tools
  • Pivoted screw cap - does not disturb dispensing
  • Easy to calibrate and adjust in order to comply with GLP guidelines
  • Easy to dismantle for cleaning
  • Replaceable filling and discharge valve with safety bulb (locks when discharge tube is not assembled)
  • 360° rotatable valve block allows bottle label to face user
  • Telescoping filling tube adjusts easily to different size bottles
  • DE-M marking
Specifications
Accuracy0.5% (125μL)
Product TypeDispenser
Angle90°
AutoclavableAutoclavable
Certifications/ComplianceCertifications: ISO 9001
ColorGray/Red
DensityUp to 2.2 g/cm3
DescriptionWith recirculation valve
Volume (Metric)2.5 to 25 mL
DisplayDigital
For Use With (Application)Dispensing aggressive reagents, including concentrated bases and acids such as H3PO4, H2SO4 (except e.g., HCl, HNO3 und HF), saline solutions, as well as many organic solvents
IncludesDispensette S bottle-top dispenser,a GL 45 threaded bottle adapter, DE-M marking, performance certificate, telescoping filling tube, recirculation valve with an optional recirculation tube, a mounting tool, and polypropylene adapters (GL 32-33, GL38, S40)
Increments0.1 mL
Precision0.1% (25μL)
Temperature (English)59°F to 104°F
Temperature (Metric)15°C to 40°C
Viscosity RatingUp to 500 mm 2/s
Unit SizeEA