Search Fisher Scientific
| Catalog Number | Capacity (Metric) | Quantity |
|---|---|---|
| FIS13-688-74 | 1.5 to 3 mL | 100 Pack |
| FIS03-840-300 | 70 to 850 μL | 100 Pack, Individually Wrapped |
| FIS13-688-73 | 1.5 to 3 mL | 500 Pack |
| FKS10566581 | 1.5 to 3 mL | 100 Pack |
| FIS13-878-121 | 70 to 850 μL | 100 Pack |
| FKS10290272 | 70 to 850 μL | 100 Pack |
| FIS13-878-123 | 70 to 550 μL | 100 Pack |
| FIS13-878-124 | 70 to 550 μL | 500 Pack |
| FIS14-377-009 | 2.5 to 4.5 mL | 100 Pack |
| FKS10046731 | 2.5 to 4.5 mL | 100 Pack |
CERTIFIED LIFE SCIENCE QUALITY cuvettes from BRAND™ are manufactured from virgin raw materials under controlled room conditions, with intensive in-process controls, visual inspections, batch management, and final product inspection to ensure proven functionality.
Ultramicro
Semimicro
Macro