Pipette Tips, 1 to 10 mL
Actual product may vary
BrandTech
Pipette Tips, 1 to 10 mL
Designed for high-quality micro-pipettes and suitable for various applications. BRAND™ 1 to 10 mL pipette tips ensure precise and efficient pipetting. Tips are cleanroom-produced, free from contaminants, and environmentally friendly.
Have Questions?
Catalog NumberPackaging
FIS13-688-654Bagged, 2 x 100 Tips
Catalog number FIS13-688-654
Price (SGD)
332.00
Pack
Packaging:
Bagged, 2 x 100 Tips
Price (SGD)
332.00
Pack
  • Engineered to fit the Transferpette™ line of micropipettes
  • Compatibility tested with other micropipettors from various manufacturers
  • Manufactured and packaged under certified cleanroom conditions according to ISO 14644-1, Class 7 and 8
  • Made from high-quality, pure raw materials, free of DiHEMDA and oleamide
  • No use of mold release agents such as stearates and erucamide in the production process
  • Thin-walled for high translucency, easy ejection, and low waste
  • Bulk tips are colorless and placed in resealable, clear bags
  • Racked tips are colorless and placed into blue-colored tip trays
  • TipBoxes are made of 100% PP and are fully recyclable
Specifications
Volume (Metric)1 to 10 mL
FilteredNon-filtered
SterilityNon-sterile
ColorColorless
AutoclavableAutoclavable
Length (Metric)15.65cm
Clean ClaimsDiHEMDA- and oleamide-free
FormatBulk
Tip StyleStandard
PackagingBagged, 2 x 100 Tips
Product TypePipette Tips
DisposableLimited-use
For Use With (Equipment)Transferpette and most Eppendorf models
GraduatedNo