BRAND™ Dispensette™ III Digital and Analog Bottle-Top Dispensers
BRAND™ Dispensette™ III Digital and Analog Bottle-Top Dispensers
Actual product may vary
Fisherbrand
BRAND™ Dispensette™ III Digital and Analog Bottle-Top Dispensers
For safe and accurate dispensing of many laboratory liquids, including most acids, bases, salt solutions, scintillation fluids and many organic solvents
Have Questions?
Catalog NumberVolume (Metric)
FIS13-688-2281 to 10 mL
Catalog number FIS13-688-228
Price (SGD)
-
Volume (Metric):
1 to 10 mL
Low-friction floating piston design provides reliable, smooth, one-handed operation without wiping seals to wear out. Suitable for most laboratory applications and fit most bottles with included adapters. Autoclavable for use with sterile reagents.
  • Digital Easy Calibration™ and analog models from 0.05 to 100mL
  • Rugged floating piston design adapts to nearly any unpressurized container
  • All models can be calibrated easily
  • Digital models have mechanical digital displays and include Easy Calibration feature
  • Integral SafetyPrime™ valve permits recirculation of dispensed liquid into reagent bottle during priming to reduce sputtering and conserve reagents
  • Safety discharge system protects against inadvertent dispensing
  • Polypropylene cylinder sleeve helps contain dispenser contents in event of breakage
  • Flowpath consists of: borosilicate glass, Al2O3 ceramic, platinum-iridium, PFTE, ETFE, FEP and PFA
  • Polypropylene safety screw cap on discharge tube
  • Accuracy for all models is within ±≤0.5% (±≤1.0% for 0.05-0.5mL size), with reproducibility of ≤0.1% (≤0.2% for 0.05-0.5mL size)
  • Autoclavable at 250°F (121°C) for sterile dispensing or decontamination
  • Supplied with thread adapters and telescoping filling tube that adjusts to length without cutting
  • Valve block rotates 360°
For dispensing concentrated fuming acids, peroxides, or difficult organic solvents, select the Dispensette Organic. Not for use with hydrofluoric acid (HF) or carbon disulfide (CS2).
Specifications
Accuracy≤0.5% ±50μL
Product TypeDispensette III Bottletop Dispenser
Angle90°
AutoclavableAutoclavable
Certifications/ComplianceGLP, ISO 9000
DensityUp to 2.2 g/cm3
MaterialBorosilicate Glass
Volume (Metric)1 to 10 mL
DisplayAnalog
IncludesSafetyPrime valve, 45mm thread, discharge tube, 125 to 240mm L telescoping filling tube, valve mounting/calibration tool, 28, 33 and 38mm thread polypropylene adapters, certificate of performance and operating manual
Increments0.2 mL
Max. Pressure600 mbar
Precision≤0.1% 10μL
Temperature (English)59°F to 104°F
Temperature (Metric)15°C to 40°C
Viscosity Rating500mm2/sec.
Warranty1 Year
Unit SizeEA