Disposable Pipette Basins
Disposable Pipette Basins
Actual product may vary
Disposable Pipette Basins
Disposable Pipette Basins
Disposable Pipette Basins
Disposable basins offer convenience, fast cleanup.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)SterilityPackaging
FIS13-681-50050 mLNon-sterileBulk
FIS13-681-50825 mLSterileIndividually Wrapped
FIS13-681-50925 mLSterileBag
FIS13-681-50150 mLSterileIndividually Wrapped
FIS13-681-50250 mLSterileBulk
Catalog number FIS13-681-500
Price (SGD)
24.00
Case
Add to cart
Capacity (Metric):
50 mL
Sterility:
Non-sterile
Packaging:
Bulk
Price (SGD)
24.00
Case
Add to cart
Fisherbrand™ Disposable Pipette Basins are packaged in easy-to-open bags to reduce contamination risks. They offer convenience and fast cleanup.
  • Easy-pour corners for quick disposal of excess sample solution
25mL Well Basins
  • Dimensions: 5.7L × 2.4W × 0.8 in.H (14.5 × 6.3 × 2.1cm)
50mL Basins
  • Angled sides and V-shaped bottom permit easy withdrawal of sample solution
  • Basin is constructed of clear PVC; Sterile (unless opened)
  • Dimensions: 5.75L × 2.75W × 1 in.H (14.0 × 7 × 2.5cm)
100mL Basins
  • Versatile design allows ample clearance for all types of 4-, 8-, and 12-channel pipetters
  • Ideal for extended multichannel pipetting into microtiter plates, tubes, and deep-well blocks
  • Dimensions: 5.75L × 3.5W × 1.25 in.H (14.6 × 9 × 3.2cm)
Specifications
ColorClear
Capacity (Metric)50 mL
Dimensions (L x W x H)5.75 x 2.75 x 1 in. (14.6 x 7 x 2.54cm)
MaterialPVC
SterilityNon-sterile
TypePipetting Reservoir
Height (English)1 in.
Height (Metric)2.54 cm
Length (English)5.75 in.
Length (Metric)14.6 cm
PackagingBulk
Width (English)2.75 in.
Width (Metric)7 cm