Squibb Pear-Shaped Separatory Funnels with PTFE-Plug Stopcock
Actual product may vary
Squibb Pear-Shaped Separatory Funnels with PTFE-Plug Stopcock
Do not require grease
Have Questions?
Catalog NumberCapacity (Metric)Height (Metric)Stopper Number
FIS13-678-605500 mL323 mm27
Catalog number FIS13-678-605
Price (SGD)
1,001.00
Case
Capacity (Metric):
500 mL
Height (Metric):
323 mm
Stopper Number:
27
Price (SGD)
1,001.00
Case
Do not require grease
Lower stems made of large-diameter tubing for liquid to drain out completely when stopcock is open.
  • KIMAX glass
  • Eliminate contamination
  • Will not bind or freeze
Specifications
Capacity (Metric)500 mL
MaterialBorosilicate Glass
Height (Metric)323 mm
Certifications/ComplianceASTM E1096, Type IV
For Use With (Application)Eliminates contamination
Length (Metric) Stem60mm
Quantity6 Case
Stopcock4; PTFE
Stopper MaterialGlass
Stopper Number27
Product TypeSquibb Separatory Funnel
Unit SizeCase