Reusable Class A Volumetric Pipets
Actual product may vary
Reusable Class A Volumetric Pipets
Available in either standard or serialized models.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)TypeMin. Flow TimeAccuracy
FIS13-650-3P25 mLSerialized Volumetric Pipet25 sec.±0.03 mL
FIS13-650-3J8 mLSerialized Volumetric Pipet10 sec.±0.02 mL
FIS13-650-2S50 mLVolumetric Pipet25 sec.±0.05 mL
FIS13-650-3B1 mLSerialized Volumetric Pipet8 sec.±0.006 mL
FIS13-650-3C2 mLSerialized Volumetric Pipet8 sec.±0.006 mL
FIS13-650-3V100 mLSerialized Volumetric Pipet30 sec.±0.08 mL
FIS13-650-2L10 mLVolumetric Pipet15 sec.±0.02 mL
FIS13-650-2W200 mLVolumetric Pipet30 sec.±0.16 mL
FIS13-650-3S50 mLSerialized Volumetric Pipet25 sec.±0.05 mL
FIS13-650-3F5 mLSerialized Volumetric Pipet8 sec.±0.01 mL
FIS13-650-2R40 mLVolumetric Pipet25 sec.±0.05 mL
FIS13-650-2B1 mLVolumetric Pipet8 sec.±0.006 mL
FIS13-650-3H7 mLSerialized Volumetric Pipet10 sec.±0.01 mL
FIS13-650-3K9 mLSerialized Volumetric Pipet10 sec.±0.02 mL
FIS13-650-2U100 mLVolumetric Pipet30 sec.±0.08 mL
FIS13-650-2K9 mLVolumetric Pipet10 sec.±0.02 mL
FIS13-650-2A0.5 mLVolumetric Pipet5 sec.±0.006 mL
FIS13-650-2T75 mLVolumetric Pipet30 sec.±0.08 mL
FIS13-650-2M15 mLVolumetric Pipet25 sec.±0.03 mL
FIS13-650-2D3 mLVolumetric Pipet8 sec.±0.01 mL
FIS13-650-2E4 mLVolumetric Pipet8 sec.±0.01 mL
FIS13-650-3R40 mLSerialized Volumetric Pipet25 sec.±0.05 mL
FIS13-650-3L10 mLSerialized Volumetric Pipet15 sec.±0.02 mL
FIS13-650-2Y2.5 mLVolumetric Pipet8 sec.±0.006 mL
FIS13-6502Y2.5 mLVolumetric Pipet8 sec.±0.006 mL
FIS13-650-2C2 mLVolumetric Pipet8 sec.±0.006 mL
FIS13-650-3Q30 mLSerialized Volumetric Pipet25 sec.±0.03 mL
FIS13-650-2P25 mLVolumetric Pipet25 sec.±0.03 mL
FIS13-650-2N20 mLVolumetric Pipet25 sec.±0.03 mL
FIS13-650-2Q30 mLVolumetric Pipet25 sec.±0.03 mL
FIS13-650-2G6 mLVolumetric Pipet10 sec.±0.01 mL
FIS13-650-2H7 mLVolumetric Pipet10 sec.±0.01 mL
FIS13-650-3D3 mLSerialized Volumetric Pipet8 sec.±0.01 mL
FIS13-650-3M15 mLSerialized Volumetric Pipet25 sec.±0.03 mL
FIS13-650-2J8 mLVolumetric Pipet10 sec.±0.02 mL
FIS13-650-3E4 mLSerialized Volumetric Pipet8 sec.±0.01 mL
FIS13-650-2F5 mLVolumetric Pipet8 sec.±0.01 mL
FIS13-650-3A0.5 mLSerialized Volumetric Pipet5 sec.±0.006 mL
FIS13-650-3N20 mLSerialized Volumetric Pipet25 sec.±0.03 mL
FIS13-650-3G6 mLSerialized Volumetric Pipet10 sec.±0.01 mL
Catalog number FIS13-650-3P
Price (SGD)
489.00
Case
Volume (Metric):
25 mL
Type:
Serialized Volumetric Pipet
Min. Flow Time:
25 sec.
Accuracy:
±0.03 mL
Price (SGD)
489.00
Case
Available in either standard or serialized models.
  • Borosilicate glass
  • Volume accuracy marked on each pipet
  • Permanent amber markings
  • Tuf-Temp™–treated tempered tips reduce chipping
Specifications
Accuracy±0.03 mL
CalibrationIndividually calibrated
Certifications/ComplianceASTM E542, ASTM E969, certificate of traceability, NIST
ClassA
MaterialBorosilicate Glass
GraduatedYes
Graduation ColorAmber
Min. Flow Time25 sec.
Tip StyleTuf-Temp™ treated
Volume (English)0.85 oz.
ColorBlue
Volume (Metric)25 mL
TypeSerialized Volumetric Pipet