Orion Star™ A121 Portable pH Meter
Orion Star™ A121 Portable pH Meter
Actual product may vary
Orion Star™ A121 Portable pH Meter
Orion Star™ A121 Portable pH Meter
Orion Star™ A121 Portable pH Meter
Orion Star™ A121 Portable pH Meter
Thermo Scientific
Orion Star™ A121 Portable pH Meter
Take simple pH and temperature measurements with our portable pH meter with easy operation for basic field testing.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
FIS13-645-506Portable pH Meter Standard Kit
FIS13-645-504Portable pH Meter
Catalog number FIS13-645-506
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Type:
Portable pH Meter Standard Kit
Take simple, routine pH, mV and temperature measurements with the Thermo Scientific™ Orion Star™ A121 Portable pH Meter. This meter is an easy-to-use and budget-friendly instrument for basic pH analysis in the field. Perform up to a 3 point pH calibration with easy recall of calibration points and slope. Quickly navigate setup menus and ensure consistent calibrations using the simple keypad layout and on-screen text prompts. This waterproof meter has an IP67-rated housing for rugged protection in outdoor environments.
Thermo Scientific Orion Star A121 Portable pH Meters are a simple, economical solution for your routine outdoor sample analysis. Featuring an emphasized measure key, selectable read modes and measurement stability indicator, our basic Orion Star A121 pH Meters are designed to deliver measurements you can trust at an affordable price.
  • Measure pH, mV or relative mV with temperature
  • Perform up to a 3 point pH calibration with automatic buffer recognition or manual buffer entry
  • Simplified, easy-to-read display shows main measurement with temperature, stability indicator and electrode condition icon
  • User-friendly operation with basic calibration text prompts
  • Easily recall calibration data for procedure checks
  • Non-volatile memory preserves 50 point internal data log and meter settings, even with loss of power
  • Flexible power options with AA batteries or optional universal power adapter
  • IP67-rated housing is waterproof for rugged field operation
  • Meter kits include hard-sided field case and protective meter armor with electrode holders, adjustable wrist/belt strap and built-in meter stand
  • Meters offer CE, TUV 3-in-1 and FCC Class A certifications
  • 3-year replacement warranty
Disclaimer to internal research use restriction:
Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Accuracy (Temperature)±0.1°C
Accuracy (mV)±0.2 or ±0.05%, whichever is greater
Accuracy (pH)±0.01
Data Management DetailsInternal Data Point Memory
DescriptionOrion Star A121 portable pH meter standard kit with general purpose pH/ATC electrode
OutputsNone
IncludesStar A121 pH meter, 9107BNMD gel-filled epoxy-body pH/ATC electrode, 916099 pH buffer kit, protective meter armor, hard-sided carrying case, 4 AA batteries
Measurement ModespH, mV, Relative mV with Temperature
CalibrationpH, relative mV (RmV) and temperature
Range (pH)-2 to 16
Resolution (pH)0.01, 0.1
Range (mV)±1600.0
Resolution (mV)0.1mV
Calibration Points1 to 3
Temperature Range (Metric)-5°C to +105°C
Temperature Range (English)22.0°F to 221.0°F
Resolution (Temperature)0.1°C, 0.1°F
Temperature SelectionManual or automatic with ATC temperature probe
Temperature Probe Calibration1 point temperature offset calibration
Display TypeLCD
MemoryNon-volatile memory preserves log and settings
Data Points50
Log Function TypeAutomatic with Auto-Read Measure Mode, Manual with Continuous Measure Mode
InputsBNC (pH or ORP electrode), 8 pin MiniDIN (ATC temperature probe)
Certifications/ComplianceCE, TUV 3-1, FCC Class A
IP RatingIP67
AC AdapterUniversal 50-60 Hz, 100-240 VAC power adapter (sold separately, Cat. No. 1010003)
Dimensions (L x W x H)24 cm x 10 cm x 6.5 cm (9.5 in. x 3.9 in. x 2.6 in.)
Weight (Metric)0.5 kg
Weight (English)2 lb.
Warranty3 Year Meter Warranty
KeypadSimple with highlighted measure key, scroll and shortcut keys
ModelOrion Star A121 Portable pH/mV/RmV/Temperature Meter
Power Supply4 AA batteries (included) or universal AC adapter (sold separately)
Probe TypepH electrode, ORP electrode, ATC temperature probe (includes 9107BNMD Orion Triode pH/ATC electrode)
LanguagesEnglish
Range (Relative mV)±1999.9mV
Relative Humidity Range5 to 85%, non-condensing
Isopotential Point7.00pH
StabilitySmart Stability and Smart Averaging
Temperature (English) Operating41°F to 113°F (ambient)
Temperature (Metric) Operating5°C to 45°C (ambient)
Channels1
Battery Type4 x AA
TypePortable pH Meter Standard Kit
Battery Life2000 hr.
Unit SizeEA