Orion™ Pure Water™ pH Buffers and pHISA™ Adjustor
Actual product may vary
Thermo Scientific
Orion™ Pure Water™ pH Buffers and pHISA™ Adjustor
Sample well, rain, distilled/deionized, boiler feed and process waters with this adjustor.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberChemical Name or Material
FIS13-641-165pH Buffer B
FIS13-641-164pH Buffer A
Catalog number FIS13-641-165
Price (SGD)
1,251.00
Pack
Chemical Name or Material:
pH Buffer B
Price (SGD)
1,251.00
Pack
Sample well water, rain water, distilled/deionized water, boiler feed water, process water and more with the Thermo Scientific™ Orion™ Pure Water™ pH Buffers and pHISA™ Adjustor. It offers fast, accurate pH results in high purity and low ionic strength waters.
Features:
  • Increases measurement accuracy
  • Reduces electrode drift
  • Eliminates contamination of sample
Pure Water pH Test Kit:
  • Kit contains special low ionic strength buffers for calibration and pHISA™ adjustor additive, to increase sample ionic strength without changing its pH
  • For best results, a high quality pH electrode such as the Thermo Scientific™ Orion™ ROSS Ultra™ glass-body pH electrode, Cat. No. 8102BNUWP, is recommended
Warranty: Out of box
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Chemical Name or MaterialpH Buffer B
Boiling Point100°C
ColorClear
For Use With (Application)Provide fast, accurate pH results in high purity and low ionic strength waters. Recommended for samples such as well water, rain water, distilled/deionized water, boiler feed water and process water.
For Use With (Equipment)pH Meter and Electrode
pH4.1
Physical FormLiquid
Quantity4 x 475 mL
PackagingBottle
Solubility InformationSoluble in Water
Unit SizePack