Glass Kinematic Viscometer Tubes
Glass Kinematic Viscometer Tubes
Actual product may vary
Glass Kinematic Viscometer Tubes
Cannon™-Fenske type viscometer tubes for measuring kinematic viscosity of petroleum products by ASTM™ D 445.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberApproximate ConstantASTM NumbersSizeDescription
FIS13-617L20 cSt/sec.600600Calibrated
FIS13-617K8 cSt/sec.500500Calibrated
FIS13-617G0.5 cSt/sec.350350Calibrated
FIS13-617H1.2 cSt/sec.400400Calibrated
FIS13-617B0.004 cSt/sec.5050Calibrated
FIS13-616F0.25 cSt/sec.300300Uncalibrated
FIS13-616A0.002 cSt/sec.2525Uncalibrated
FIS13-616H1.2 cSt/sec.400400Uncalibrated
FIS13-616D0.035 cSt/sec.150150Uncalibrated
FIS13-617E0.1 cSt/sec.200200Calibrated
FIS13-616M0.008 cSt/sec.7575Uncalibrated
FIS13-617A0.002 cSt/sec.2525Calibrated
FIS13-616C0.015 cSt/sec.100100Uncalibrated
FIS13-616E0.1 cSt/sec.200200Uncalibrated
FIS13-617D0.035 cSt/sec.150150Calibrated
FIS13-616K8 cSt/sec.500500Uncalibrated
FIS13-617J2.5 cSt/sec.450450Calibrated
FIS13-617F0.25 cSt/sec.300300Calibrated
FIS13-616G0.5 cSt/sec.350350Uncalibrated
Catalog number FIS13-617L
Price (SGD)
665.00
EA
Add to cart
Approximate Constant:
20 cSt/sec.
ASTM Numbers:
600
Size:
600
Description:
Calibrated
Price (SGD)
665.00
EA
Add to cart
Cannon™-Fenske type viscometer tubes for measuring kinematic viscosity of petroleum products by ASTM™ D 445.
  • Available calibrated (with calibration certificate) or uncalibrated
  • Excellent precision
  • Borosilicate glass
  • Specifications also conform with ASTM D 446
  • Minimum sample volume, 7mL
  • Requires liquid bath with depth of 203mm
Specifications
Product TypeCannon-Fenske Routine Viscometer
Viscosity Range4000 to 20,000 cSt
CalibratedYes
DescriptionCalibrated
Approximate Constant20 cSt/sec.
ASTM Numbers600
IncludesInstruction sheet, calibration certificate
MaterialBorosilicate Glass
Depth (English) Bath8 in.
Depth (Metric) Bath20.3 cm
AccuracyASTM D445, ASTM D446, ISO 3104, ISO 3105
Precision±0.2%
For Use With (Application)For measuring kinematic viscosity of transparent Newtonian liquids, particularly petroleum products or lubricants
Size600
Volume (Metric)7 mL
Unit SizeEA