Glass Ubbelohde Calibrated Viscometer Tubes
Glass Ubbelohde Calibrated Viscometer Tubes
Actual product may vary
Glass Ubbelohde Calibrated Viscometer Tubes
For measurement of kinematic viscosities of petroleum products according to ASTM™ D 445 and D 446.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberApproximate ConstantASTM NumbersSizeViscosity Range
FIS13-614J10 cSt/sec.442000 to 10,000 cSt
FIS13-614A0.01 cSt/sec.112 to 10 cSt
FIS13-614C0.05 cSt/sec.1B1B10 to 50 cSt
FIS13-614E0.3 cSt/sec.2C2C60 to 300 cSt
FIS13-614G0.5 cSt/sec.2B2B100 to 500 cSt
FIS13-614H3 cSt/sec.3C3C600 to 3000 cSt
FIS13-614F0.003 cSt/sec.0C0C0.6 to 3 cSt
Catalog number FIS13-614J
Price (SGD)
967.00
EA
Add to cart
Approximate Constant:
10 cSt/sec.
ASTM Numbers:
4
Size:
4
Viscosity Range:
2000 to 10,000 cSt
Price (SGD)
967.00
EA
Add to cart
For measurement of kinematic viscosities of petroleum products according to ASTM™ D 445 and D 446.
  • Certified accurate (certificate included)
  • Excellent precision
  • Borosilicate glass
  • Minimum sample volume: 11mL
  • Require liquid bath depth of 241mm
Specifications
Product TypeUbbelohde Viscometer
Viscosity Range2000 to 10,000 cSt
CalibratedYes
Approximate Constant10 cSt/sec.
ASTM Numbers4
Certifications/ComplianceASTM D445, ASTM D446, ISO 3104, ISO 3105, calibration certificate
IncludesInstruction sheet, calibration certificate
MaterialBorosilicate Glass
Depth (English) Bath9.5 in.
Depth (Metric) Bath24.1 cm
AccuracyASTM D445, ASTM D446
Precision±0.2%
For Use With (Application)For measurement of kinematic viscosity of transparent Newtonian liquids
Size4
Volume (Metric)11 mL
Unit SizeEA