Parafilm™ M Laboratory Film
Actual product may vary
Fisherbrand
Parafilm™ M Laboratory Film
Efficient and cost-effective covering for test tubes, beakers, vials, petri dishes, flasks, and various other instruments and objects with irregular surfaces.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Width (Metric)Color
FIS13-374-1276 m10 cmNatural
FIS13-374-1676 m5 cmNatural
FIS13-374-1038 m10 cmNatural
FKS1177264438 m10 cmNatural
Catalog number FIS13-374-12
Price (SGD)
80.60
Special offer
Ends: 06-Sep-2026
124.00
Save 43.40 (35%)
Each
Length (Metric):
76 m
Width (Metric):
10 cm
Color:
Natural
Price (SGD)
80.60
Special offer
Ends: 06-Sep-2026
124.00
Save 43.40 (35%)
Each
Efficient and cost-effective covering for test tubes, beakers, vials, petri dishes, flasks, and various other instruments and objects with irregular surfaces.
  • Protect the integrity of research applications with a tight moisture-barrier that holds a b seal and doesn't release from glassware, despite off-gassing
  • Reduce the risk of tampered data with a covering that is odorless, colorless and semi-transparent
  • Prevents moisture and volume loss due to material composition and tight seal
  • Flexible and self-sealing material allows it to cling around irregular shapes and surfaces
  • Stretches more than 200% of original length
  • Resistant up to 48 hours against many polar substances such as saline solutions, inorganic acids and alkaline solutions
Permeability
  • Oxygen: (ASTM 1927-98) 150 cc/m2 d at 23°C and 50% RH
  • Carbon Dioxide: (Modulated IR Method) 1200 cc/m2 d at 23°C and 0% RH
  • Water Vapor: (ASTM F1249-01)
    • Flat: 1 g/m2 d at 38°C and 90% RH
    • Creased: 1 g/m2 d at 38°C and 90% RH
Effects of Common Reagents
  • Potassium Permanganate
    • 5%: No apparent effect except permanent dark brown coloration in 18 hours
    • 0.1%: Same as 5% except slightly less color
  • Iodine solution (0.1N)
    • No effect except staining brown in 18 hours
  • Ethyl Alcohol (95%)
    • No apparent effect except some face whitening in 24 hours
  • No apparent effect in 24 hours:
    • Hydrochloric Acid conc. (12N) dil. (5N)
    • Sulphuric Acid conc. (36N) dil. (5N)
    • Nitric Acid conc. (16N) dil. (5N)
    • Sodium Hydroxide conc. (22%)
    • Ammonium Hydroxide conc. (28% NH3)
    • Salt (NaCl) solution (20%)
    • Isopropyl Alcohol (99%)
  • Not recommended for use with chlorinated, non-polar aliphatic and aromatic solvents
  • Film becomes soft and sticky at about 130° to 150°F (54° to 66°C)
Specifications
ApplicationsEfficient and cost-effective covering, over wrap to further secure stoppers, lids, and caps, and shelf or tray liner to prevent slipping of bottles or instruments.
Length (English)250 ft.
ColorNatural
DescriptionAmcor PM999
Item DescriptionSize: 4 in x 250 ft./roll
Product TypeFilm
Width (English)4 in.
Width (Metric)10 cm
Length (Metric)76 m
MaterialThermoplastic
Thickness5 mil
Unit SizeEach