▸
Racks
▸
Vial Racks
▸
COD Vial Rack
Actual product may vary
CHEMetrics
COD Vial Rack
Holds 40 vials
Have Questions?
Catalog Number
Holds
FIS13-299-225
Vials
Catalog number
FIS13-299-225
Price (SGD)
247.00
Each
Add to cart
Holds:
Vials
Price (SGD)
247.00
Each
Add to cart
Stainless steel
Perforated holds bent to form the rack
Four rows of 10
Specifications
Autoclavable
Not Autoclavable
Holds
Vials
Material
Stainless Steel
Positions
40
Product Type
COD Vial Rack