CHEMets Phenol Test Kit Refill
Actual product may vary
CHEMetrics
CHEMets Phenol Test Kit Refill
Provides results in two minutes or less
Have Questions?
Catalog NumberNo. of Tests
FIS13-299-205A30
Catalog number FIS13-299-205A
Price (SGD)
124.00
Pack
No. of Tests:
30
Price (SGD)
124.00
Pack
This product contains 30 self-filling CHEMets ampules to refill kit K-8012 for the analysis of phenols. R-8012 is compatible with C-8001 and C-8012 comparators.
Specifications
AnalytesPhenol
For Use With (Equipment)C-8001 and C-8012 comparators
Includes30 self-filling CHEMets ampoules to refill kit K-8012
No. of Tests30
Range0 to 1 ppm and 1 to 12 ppm
Shelf Life1 year
Test Method4-Aminoantipyrine
TypeCHEMets Refill Kit
Unit SizePack